Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009208.3 | NP_033234.2 | 6 | 992 | |
GenBank mRNA | AK045583.1 | - | 5 | 879 | |
AK147218.1 | - | 1004 | |||
BC066086.1 | - | 7 | 851 | ||
BC080271.1 | - | 6 | 987 | 6831942 | |
M28383.1 | - | 6 | 962 |