Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177721.4 | NP_808389.2 | 1 | 2321 | |
GenBank mRNA | AK083089.1 | - | 1 | 301 | |
AK161331.1 | - | 1 | 23 | ||
BC026487.1 | - | 1 | 223 | ||
BC056759.1 | - | 1 | 2300 | 6518485 | |
BC139395.1 | - | 1 | 2321 | 8862417 | |
BC139396.1 | - | 1 | 2321 | 8862418 |