Gene Transcripts

Gene Symbol
Melk
Entrez Gene ID
17279
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
AI327312, MPK38, mKIAA0175
Chromosome Location
Chr. 4: 44300876 - 44364675 on Build GRCm38
UniGene ID
Mm.268668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010790.2 NP_034920.2 7 728
GenBank mRNA AK010498.1 - 7 694
AK011932.1 - 7 728
AK129076.1 - 7 681
AK137000.1 - 7 754
AK145021.1 - 7 663
AK145316.1 - 7 637
AK161029.1 - 7 737
AK164138.1 - 7 695
AK167457.1 - 7 652
BC020416.1 - 6 563
BC085276.1 - 7 761 30443236
L76158.1 - 7 659
X95351.1 - 7 666

View Full Product Details