Gene Transcripts
- Gene Symbol
- PDIA3
- Entrez Gene ID
- 2923
- Gene Name
- protein disulfide isomerase family A member 3
- Gene Aliases
- ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
- Chromosome Location
- Chr. 15: 43746392 - 43772606 on Build GRCh38
- UniGene ID
- Hs.591095
- Species
- Homo sapiens
- Gene Symbol
- PDIA3P1
- Entrez Gene ID
- 171423
- Gene Name
- protein disulfide isomerase family A member 3 pseudogene 1
- Gene Aliases
- ERp60, GRP58P, PDIA3P
- Chromosome Location
- Chr. 1: 147177851 - 147179949 on Build GRCh38
- UniGene ID
- Hs.658566
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002305.1 |
|
1 |
497 |
|
GenBank mRNA |
AK295985.1 |
- |
1 |
382 |
|
BM555251.1 |
- |
1 |
441 |
|
View Full Product Details