Gene Transcripts

Gene Symbol
Ublcp1
Entrez Gene ID
79560
Gene Name
ubiquitin-like domain containing CTD phosphatase 1
Gene Aliases
4930527B16Rik, 8430435I17Rik, BC002236
Chromosome Location
Chr. 11: 44454571 - 44470548 on Build GRCm38
UniGene ID
Mm.338784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024475.5 NP_077795.2 11 1121
GenBank mRNA AK043952.1 - 11 1028
AK050031.1 - 11 1032
AK147744.1 - 11 1069
AK153998.1 - 11 1033
BC002236.1 - 4 395
BC056652.1 - 7 622
BC083331.1 - 9 782
BC085111.1 - 10 1024 30735741
BC094051.1 - 4 1175
BC106093.1 - 10 998 5098953

View Full Product Details