Gene Transcripts

Gene Symbol
Lztfl1
Entrez Gene ID
93730
Gene Name
leucine zipper transcription factor-like 1
Gene Aliases
5530402H04Rik, 6130400H19Rik, AI414725, AW048545
Chromosome Location
Chr. 9: 123694413 - 123717668 on Build GRCm38
UniGene ID
Mm.378263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033322.2 NP_201579.1 9 842
GenBank mRNA AJ297352.1 - 9 838
AK030068.1 - 9 854
AK041091.1 - 9 875
AK046515.1 - 9 831
AK054003.1 - 9 848
AK081529.1 - 7 627
AK145263.1 - 9 851
AK147312.1 - 8 786
AK157770.1 - 9 849
BC026840.1 - 9 842 4165152
BC037045.1 - 9 842
BC098224.1 - 9 847 1263280

View Full Product Details