Gene Transcripts

Gene Symbol
Reg4
Entrez Gene ID
67709
Gene Name
regenerating islet-derived family, member 4
Gene Aliases
2010002L15Rik, GISP, RELP
Chromosome Location
Chr. 3: 98222156 - 98236748 on Build GRCm38
UniGene ID
Mm.46306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026328.2 NP_080604.2 6 947
GenBank mRNA AK008049.1 - 6 949
AK008438.1 - 6 950
AK160329.1 - 6 945
AK162316.1 - 6 948

View Full Product Details