Gene Transcripts

Gene Symbol
Spcs1
Entrez Gene ID
69019
Gene Name
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases
1810004F21Rik
Chromosome Location
Chr. 14: 30999826 - 31001666 on Build GRCm38
UniGene ID
Mm.28852
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026911.3 NP_081187.2 4 809
GenBank mRNA AK007336.1 - 4 507
AK082928.1 - 4 508
AK088658.1 - 2 1047
BC020050.1 - 1165
BC034270.2 - 4 684
BC049895.1 - 4 500 3991764

View Full Product Details