Gene Transcripts

Gene Symbol
Pigo
Entrez Gene ID
56703
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class O
Gene Aliases
PIG-O
Chromosome Location
Chr. 4: 43017637 - 43027696 on Build GRCm38
UniGene ID
Mm.143738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB038560.1 - 2 1206
AK220315.1 - 2 1154
BC036977.1 - 3 551
BC092394.1 - 2 1224 30534262
BC141058.1 - 2 1162 9055670

View Full Product Details