Gene Transcripts

Gene Symbol
U2AF1
Entrez Gene ID
7307
Gene Name
U2 small nuclear RNA auxiliary factor 1
Gene Aliases
FP793, RN, RNU2AF1, U2AF35, U2AFBP
Chromosome Location
Chr. 21: 43092956 - 43108291 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025203.1 NP_001020374.1 4, 5 344
NM_001025204.1 NP_001020375.1 5, 6 411
NM_006758.2 NP_006749.1 4, 5 344
GenBank mRNA AB451244.1 - 4, 5 260
AF370386.1 - 3, 4 1067
AJ627978.1 - 4, 5 290
AK098266.1 - 1 867
AK307560.1 - 4, 5 293
BC001177.1 - 4, 5 288 3534272
BC001923.1 - 4, 5 288 2823296
BC005915.1 - 5, 6 357 4295037
CB110007.1 - 4, 5 344
M96982.1 - 4, 5 298

Gene Symbol
U2AF1L5
Entrez Gene ID
102724594
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 5
Gene Aliases
Chromosome Location
Chr. 21: 6484623 - 6499284 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320646.1 NP_001307575.1 4, 5 372
NM_001320648.1 NP_001307577.1 4, 5 372
NM_001320650.1 NP_001307579.1 4, 5 343
NM_001320651.1 NP_001307580.1 5, 6 439
GenBank mRNA AL522322.3 - 4, 5 289
BG612658.1 - 4, 5 308
CN280267.1 - 5, 6 316
CN280269.1 - 4, 5 309

View Full Product Details