Gene Transcripts

Gene Symbol
Mapkbp1
Entrez Gene ID
26390
Gene Name
mitogen-activated protein kinase binding protein 1
Gene Aliases
2810483F24Rik, AW123212, JNKBP-1, Jnkbp1, mKIAA0596
Chromosome Location
Chr. 2: 119972388 - 120027403 on Build GRCm38
UniGene ID
Mm.330501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011941.3 NP_036071.3 9 1438
GenBank mRNA AB029482.1 - 10 1252
BC070449.1 - 9 1438 30543913

View Full Product Details