Gene Transcripts
- Gene Symbol
- St8sia3
- Entrez Gene ID
- 25547
- Gene Name
- ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
- Gene Aliases
- Siat8c
- Chromosome Location
- Chr. 18: 59748766 - 59755257 on Build Rnor_6.0
- UniGene ID
- Rn.90977
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013029.1 |
NP_037161.1
|
2 |
268 |
|
GenBank mRNA |
U55938.1 |
- |
2 |
268 |
|
View Full Product Details