Gene Transcripts

Gene Symbol
OR2B6
Entrez Gene ID
26212
Gene Name
olfactory receptor family 2 subfamily B member 6
Gene Aliases
OR2B1, OR2B1P, OR2B5, OR2B6P, OR5-40, OR5-41, OR6-31, dJ408B20.2
Chromosome Location
Chr. 6: 27957241 - 27958182 on Build GRCh38
UniGene ID
Hs.532145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012367.1 NP_036499.1 1 31
GenBank mRNA BC109251.1 - 1 31
BC110457.2 - 1 31 40022996

View Full Product Details