Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243202.1 | NP_001230131.1 | 2 | 369 | |
NM_001243203.1 | NP_001230132.1 | 2 | 310 | ||
NM_012416.3 | NP_036548.1 | 1 | 1425 | ||
XM_017014624.1 | XP_016870113.1 | 2 | 330 | ||
GenBank mRNA | AF039023.1 | - | 1 | 1394 | |
AK300408.1 | - | 2 | 284 | ||
AK307840.1 | - | 1 | 1400 | ||
BC098406.1 | - | 1 | 1400 | 30347107 | |
BX537405.1 | - | 1 | 1404 | ||
DA234653.1 | - | 2 | 354 | ||
DC342914.1 | - | 2 | 294 |