Gene Transcripts

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
Chromosome Location
Chr. 16: 71644924 - 71724701 on Build GRCh38
UniGene ID
Hs.709458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289003.1 NP_001275932.1 16 3059
NM_015020.3 NP_055835.2 17 2913
GenBank mRNA AB023148.2 - 16 3059
AK001854.1 - 5 622
BC035267.1 - 12 1748
BC129927.1 - 16 2540 40124720
BX647823.1 - 17 2914
BX648157.1 - 11 1649

View Full Product Details