Gene Transcripts
- Gene Symbol
- TMEM63A
- Entrez Gene ID
- 9725
- Gene Name
- transmembrane protein 63A
- Gene Aliases
- KIAA0792
- Chromosome Location
- Chr. 1: 225840556 - 225882720 on Build GRCh38
- UniGene ID
- Hs.119387
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014698.2 |
NP_055513.2
|
11, 12 |
1452 |
|
XM_006711841.3 |
XP_006711904.1
|
9, 10 |
945 |
|
XM_011544328.2 |
XP_011542630.1
|
11, 12 |
1304 |
|
XM_011544329.2 |
XP_011542631.1
|
11, 12 |
2571 |
|
XM_011544330.2 |
XP_011542632.1
|
10, 11 |
1261 |
|
XM_011544331.2 |
XP_011542633.1
|
11, 12 |
1135 |
|
XM_011544332.2 |
XP_011542634.1
|
10, 11 |
1083 |
|
XM_017002916.1 |
XP_016858405.1
|
10, 11 |
1535 |
|
GenBank mRNA |
AB007958.1 |
- |
2, 3 |
2597 |
|
AB018335.1 |
- |
11, 12 |
1080 |
|
AK022502.1 |
- |
11, 12 |
1101 |
|
AK222934.1 |
- |
11, 12 |
1082 |
|
AK222944.1 |
- |
11, 12 |
1082 |
|
BC030245.1 |
- |
11, 12 |
1085 |
5216414 |
BC113811.1 |
- |
4, 5 |
493 |
|
View Full Product Details