Gene Transcripts
- Gene Symbol
- ASXL1
- Entrez Gene ID
- 171023
- Gene Name
- additional sex combs like 1, transcriptional regulator
- Gene Aliases
- BOPS, MDS
- Chromosome Location
- Chr. 20: 32358062 - 32439319 on Build GRCh38
- UniGene ID
- Hs.374043
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164603.1 |
NP_001158075.1
|
2, 3 |
564 |
|
NM_015338.5 |
NP_056153.2
|
2, 3 |
564 |
|
XM_006723728.3 |
XP_006723791.1
|
2, 3 |
173 |
|
XM_006723730.3 |
XP_006723793.1
|
2, 3 |
369 |
|
XM_006723732.2 |
XP_006723795.1
|
2, 3 |
345 |
|
XM_011528652.2 |
XP_011526954.1
|
3, 4 |
1506 |
|
XM_017027704.1 |
XP_016883193.1
|
2, 3 |
1629 |
|
XM_017027705.1 |
XP_016883194.1
|
2, 3 |
642 |
|
XM_017027706.1 |
XP_016883195.1
|
2, 3 |
846 |
|
GenBank mRNA |
AJ438952.2 |
- |
2, 3 |
390 |
|
AK122923.1 |
- |
2, 3 |
345 |
|
BC100280.1 |
- |
2, 3 |
558 |
|
BC137278.1 |
- |
3, 4 |
327 |
9021275 |
BC137280.1 |
- |
3, 4 |
327 |
9021277 |
BQ064719.1 |
- |
|
556 |
|
CR977683.1 |
- |
2, 3 |
564 |
|
View Full Product Details