Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016354.3 | NP_057438.3 | 3 | 1063 | |
XM_005260203.2 | XP_005260260.1 | 3 | 1060 | ||
XM_011528792.2 | XP_011527094.1 | 3 | 1060 | ||
XM_017027826.1 | XP_016883315.1 | 3 | 1044 | ||
XM_017027827.1 | XP_016883316.1 | 3 | 1060 | ||
GenBank mRNA | AB031051.1 | - | 2 | 949 | |
AF104334.1 | - | 3 | 995 | ||
AF187817.1 | - | 2 | 858 | ||
AF205072.1 | - | 3 | 1003 | ||
BC015727.1 | - | 3 | 1021 | 4908245 |