Gene Transcripts

Gene Symbol
Sipa1l3
Entrez Gene ID
292771
Gene Name
signal-induced proliferation-associated 1 like 3
Gene Aliases
Chromosome Location
Chr. 1: 87260832 - 87345368 on Build Rnor_6.0
UniGene ID
Rn.16517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013066.1 NP_001013084.1 10 3464
XM_008759130.1 XP_008757352.1 20 6891
XM_008759131.1 XP_008757353.1 20 6888
GenBank mRNA BC091303.1 - 10 3464

View Full Product Details