Gene Transcripts

Gene Symbol
NAA30
Entrez Gene ID
122830
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
C14orf35, MAK3, Mak3p, NAT12, NAT12P
Chromosome Location
Chr. 14: 57390553 - 57412748 on Build GRCh38
UniGene ID
Hs.165465
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011713.2 NP_001011713.2 5 2480
GenBank mRNA AK092674.1 - 5 2480
BC048983.1 - 1 1249
BX648700.1 - 4 1761

View Full Product Details