Gene Transcripts

Gene Symbol
Ccdc90b
Entrez Gene ID
308820
Gene Name
coiled-coil domain containing 90B
Gene Aliases
RGD1308637
Chromosome Location
Chr. 1: 157403595 - 157417153 on Build Rnor_6.0
UniGene ID
Rn.160643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024885.1 NP_001020056.1 9 1292
GenBank mRNA BC097480.1 - 9 1292
FQ215669.1 - 9 994
FQ216256.1 - 9 995
FQ218235.1 - 9 1007

View Full Product Details