Gene Transcripts
- Gene Symbol
- TARP
- Entrez Gene ID
- 445347
- Gene Name
- TCR gamma alternate reading frame protein
- Gene Aliases
- CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
- Chromosome Location
- Chr. 7: 38239577 - 38276741 on Build GRCh38
- UniGene ID
- Hs.534032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M30894.1 |
- |
2 |
420 |
|
X06774.1 |
- |
2 |
395 |
|
X06776.1 |
- |
1 |
155 |
|
View Full Product Details