Gene Transcripts

Gene Symbol
U2AF1
Entrez Gene ID
7307
Gene Name
U2 small nuclear RNA auxiliary factor 1
Gene Aliases
FP793, RN, RNU2AF1, U2AF35, U2AFBP
Chromosome Location
Chr. 21: 43092956 - 43108291 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025203.1 NP_001020374.1 8 820
NM_001025204.1 NP_001020375.1 9 887
NM_006758.2 NP_006749.1 8 820
XM_011529743.2 XP_011528045.1 7 1434
XM_017028468.1 XP_016883957.1 7 1434
GenBank mRNA AF370386.1 - 7 1543
AJ627978.1 - 8 766
AK098266.1 - 2 3213
BC001177.1 - 8 764 3534272
BC001923.1 - 8 764 2823296
BC005915.1 - 9 833 4295037
M96982.1 - 8 774

Gene Symbol
U2AF1L5
Entrez Gene ID
102724594
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 5
Gene Aliases
Chromosome Location
Chr. 21: 6484623 - 6499284 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320646.1 NP_001307575.1 8 848
NM_001320648.1 NP_001307577.1 8 848
NM_001320650.1 NP_001307579.1 8 819
NM_001320651.1 NP_001307580.1 9 915
XM_017028219.1 XP_016883708.1 7 749
XM_017028220.1 XP_016883709.1 7 749

View Full Product Details