Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021937.4 | NP_068756.2 | 5 | 1463 | |
XM_005247696.2 | XP_005247753.1 | 4 | 1299 | ||
XM_011513066.2 | XP_011511368.1 | 3 | 911 | ||
XM_017006975.1 | XP_016862464.1 | 5 | 1420 | ||
XM_017006976.1 | XP_016862465.1 | 5 | 1420 | ||
XM_017006977.1 | XP_016862466.1 | 5 | 1420 | ||
XM_017006978.1 | XP_016862467.1 | 6 | 1243 | ||
XM_017006979.1 | XP_016862468.1 | 5 | 1732 | ||
XM_017006980.1 | XP_016862469.1 | 5 | 1464 | ||
XM_017006981.1 | XP_016862470.1 | 2 | 1258 | ||
GenBank mRNA | AF268872.1 | - | 5 | 1202 | |
BC007933.2 | - | 5 | 1341 | 4120858 |