Gene Transcripts
- Gene Symbol
- RAB37
- Entrez Gene ID
- 326624
- Gene Name
- RAB37, member RAS oncogene family
- Gene Aliases
- Chromosome Location
- Chr. 17: 74671117 - 74747335 on Build GRCh38
- UniGene ID
- Hs.351413
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006638.2 |
NP_001006639.1
|
4, 5 |
376 |
|
NM_001163989.1 |
NP_001157461.1
|
4, 5 |
525 |
|
NM_001163990.1 |
NP_001157462.1
|
3, 4 |
265 |
|
NM_175738.4 |
NP_783865.1
|
4, 5 |
769 |
|
XM_005257288.4 |
XP_005257345.1
|
4, 5 |
394 |
|
GenBank mRNA |
AK054846.1 |
- |
4, 5 |
706 |
|
AK098068.1 |
- |
4, 5 |
376 |
|
AK127073.1 |
- |
4, 5 |
1308 |
|
AK290202.1 |
- |
4, 5 |
745 |
|
AK296062.1 |
- |
4, 5 |
364 |
|
AK296172.1 |
- |
3, 4 |
228 |
|
AK298033.1 |
- |
3, 4 |
261 |
|
AK303442.1 |
- |
4, 5 |
428 |
|
BC016615.1 |
- |
4, 5 |
322 |
4520191 |
BC040547.1 |
- |
4, 5 |
755 |
5287038 |
View Full Product Details