Gene Transcripts
- Gene Symbol
- Uncx
- Entrez Gene ID
- 29375
- Gene Name
- UNC homeobox
- Gene Aliases
- Chx4, PHD1, Uncx4.1
- Chromosome Location
- Chr. 12: 17182158 - 17186679 on Build Rnor_6.0
- UniGene ID
- Rn.10710
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017179.1 |
NP_058875.1
|
2 |
886 |
|
GenBank mRNA |
D87748.1 |
- |
2 |
886 |
|
View Full Product Details