Gene Transcripts

Gene Symbol
Phlpp2
Entrez Gene ID
244650
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
AI481772, C130044A18Rik, Phlppl
Chromosome Location
Chr. 8: 109868519 - 109944671 on Build GRCm38
UniGene ID
Mm.23494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122594.2 NP_001116066.2 19 6140
GenBank mRNA AK146878.1 - 1 1311
BC006885.1 - 1 274
BC030917.1 - 1 377

View Full Product Details