Gene Transcripts
- Gene Symbol
- Tacr2
- Entrez Gene ID
- 25007
- Gene Name
- tachykinin receptor 2
- Gene Aliases
- Tac2r
- Chromosome Location
- Chr. 20: 31892515 - 31905153 on Build Rnor_6.0
- UniGene ID
- Rn.202846
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080768.1 |
NP_542946.1
|
1 |
604 |
|
GenBank mRNA |
M31838.1 |
- |
1 |
604 |
|
View Full Product Details