Gene Transcripts

Gene Symbol
Slco6b1
Entrez Gene ID
170925
Gene Name
solute carrier organic anion transporter family, member 6b1
Gene Aliases
GST-1, Tst1
Chromosome Location
Chr. 9: 104388232 - 104468001 on Build Rnor_6.0
UniGene ID
Rn.27679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133412.1 NP_596903.1 1 309
XM_006245576.2 XP_006245638.1 1 379
GenBank mRNA AF321415.1 - 1 309

Gene Symbol
LOC102554121
Entrez Gene ID
102554121
Gene Name
solute carrier organic anion transporter family member 6A1-like
Gene Aliases
Chromosome Location
Chr. 9: 105246035 - 105292785 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245586.2 XP_006245648.1 1 380

View Full Product Details