Gene Transcripts
- Gene Symbol
- Rab23
- Entrez Gene ID
- 19335
- Gene Name
- RAB23, member RAS oncogene family
- Gene Aliases
- AW545388, opb, opb2
- Chromosome Location
- Chr. 1: 33719896 - 33742564 on Build GRCm38
- UniGene ID
- Mm.86744
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159729.1 |
NP_001153201.1
|
3, 4 |
487 |
|
NM_008999.4 |
NP_033025.2
|
3, 4 |
599 |
|
GenBank mRNA |
AB232618.1 |
- |
1, 2 |
164 |
|
AK016493.1 |
- |
3, 4 |
499 |
|
AK045454.1 |
- |
3, 4 |
358 |
|
AK047068.1 |
- |
3, 4 |
600 |
|
AK082017.1 |
- |
3, 4 |
562 |
|
AK163909.1 |
- |
3, 4 |
608 |
|
AK164057.1 |
- |
3, 4 |
683 |
|
M79305.1 |
- |
1, 2 |
104 |
|
Z22821.1 |
- |
1, 2 |
184 |
|
View Full Product Details