Gene Transcripts

Gene Symbol
Timm10
Entrez Gene ID
30059
Gene Name
translocase of inner mitochondrial membrane 10
Gene Aliases
Tim13, Timm13a
Chromosome Location
Chr. 2: 84827021 - 84830213 on Build GRCm38
UniGene ID
Mm.21826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013899.2 NP_038927.2 3 296
GenBank mRNA AF150090.1 - 3 273
AK008088.1 - 3 297
AK013546.1 - 3 228
BC031448.1 - 3 274 3257567

View Full Product Details