Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019537.2 | NP_062410.1 | 5 | 513 | |
GenBank mRNA | AJ238270.1 | - | 5 | 515 | |
AK042203.1 | - | 5 | 524 | ||
AK088118.1 | - | 5 | 526 | ||
BC032965.1 | - | 5 | 506 | 1196077 | |
BC060285.1 | - | 5 | 527 | 6508084 |