Gene Transcripts

Gene Symbol
Csnk1a1
Entrez Gene ID
93687
Gene Name
casein kinase 1, alpha 1
Gene Aliases
2610208K14Rik, 4632404G05Rik, 5430427P18Rik, CK1a, Csnk1a
Chromosome Location
Chr. 18: 61555165 - 61590315 on Build GRCm38
UniGene ID
Mm.26908
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146087.2 NP_666199.1 1, 2 300
GenBank mRNA AK019176.1 - 1, 2 548
AK076070.1 - 1, 2 567
AK076264.1 - 1, 2 561
AK078398.1 - 1, 2 549
AK141421.1 - 1, 2 554
AK146873.1 - 1, 2 570
AK147165.1 - 1, 2 545
AK152387.1 - 1, 2 545
AK152793.1 - 1, 2 565
AK166429.1 - 1, 2 541
AK167550.1 - 1, 2 569
AK210280.1 - 1, 2 71
BC019740.1 - 1, 2 300 3487913
BC025439.1 - 1, 2 247 5038433
BC048081.1 - 1, 2 333 6309504

View Full Product Details