Gene Transcripts

Gene Symbol
Sh2d7
Entrez Gene ID
244885
Gene Name
SH2 domain containing 7
Gene Aliases
4933412E14Rik
Chromosome Location
Chr. 9: 54534140 - 54545030 on Build GRCm38
UniGene ID
Mm.358924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173778.3 NP_776139.1 3 560
GenBank mRNA AK030178.1 - 3 561
BC116842.1 - 3 483 40126161
BC116844.1 - 3 483 40126163

View Full Product Details