Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025105.2 | NP_001020276.1 | 3 | 881 | |
NM_001271741.1 | NP_001258670.1 | 3 | 881 | ||
NM_001271742.1 | NP_001258671.1 | 2 | 320 | ||
NM_001892.5 | NP_001883.4 | 3 | 881 | ||
GenBank mRNA | AF218004.1 | - | 3 | 679 | |
AK293761.1 | - | 2 | 287 | ||
AK294942.1 | - | 4 | 468 | ||
BC007246.1 | - | 3 | 702 | ||
BC008717.2 | - | 3 | 423 | 2822250 | |
BC021971.2 | - | 3 | 700 | ||
BC025371.2 | - | 3 | 700 | ||
BT019829.1 | - | 3 | 293 | ||
BT019830.1 | - | 3 | 293 | ||
CR542206.1 | - | 3 | 293 | ||
DQ082865.1 | - | 3 | 293 | ||
EU668329.1 | - | 3 | 293 | ||
HY119498.1 | - | 2 | 320 | ||
L37042.1 | - | 3 | 504 | ||
X80693.1 | - | 3 | 418 |