Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015697.7 | NP_056512.5 | 7 | 1129 | |
XM_017008031.1 | XP_016863520.1 | 7 | 1252 | ||
GenBank mRNA | AF091086.1 | - | 7 | 797 | |
AJ621061.3 | - | 7 | 1129 | ||
AK129583.1 | - | 7 | 885 | ||
BC008804.2 | - | 7 | 986 | 3954787 | |
BC020728.1 | - | 7 | 1026 | 4696019 | |
BC116454.1 | - | 2 | 173 | ||
BM126596.1 | - | 1 | 525 | ||
BQ363759.1 | - | 4 | 405 |