Gene Transcripts

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
Chromosome Location
Chr. 16: 53491028 - 53504411 on Build GRCh38
UniGene ID
Hs.380897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012398.2 NP_001012398.1 7 792
NM_001308325.1 NP_001295254.1 7 763
NM_022476.3 NP_071921.1 7 763
XM_005256095.4 XP_005256152.1 7 1112
XM_005256096.4 XP_005256153.1 7 690
XM_005256097.4 XP_005256154.1 7 724
XM_005256098.4 XP_005256155.1 7 688
XM_017023564.1 XP_016879053.1 7 1112
XM_017023565.1 XP_016879054.1 7 690
XM_017023566.1 XP_016879055.1 7 688
GenBank mRNA AK023320.1 - 7 698
AK222743.1 - 7 695
BC001134.2 - 7 711 2967019
BC095401.1 - 7 685 30334599
CR457308.1 - 6 535

View Full Product Details