Gene Transcripts
- Gene Symbol
- REG4
- Entrez Gene ID
- 83998
- Gene Name
- regenerating family member 4
- Gene Aliases
- GISP, REG-IV, RELP
- Chromosome Location
- Chr. 1: 119794018 - 119811580 on Build GRCh38
- UniGene ID
- Hs.660883
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159352.1 |
NP_001152824.1
|
3, 4 |
502 |
|
NM_001159353.1 |
NP_001152825.1
|
2, 3 |
328 |
|
NM_032044.3 |
NP_114433.1
|
2, 3 |
328 |
|
GenBank mRNA |
AF254415.1 |
- |
2, 3 |
243 |
|
AF345934.1 |
- |
2, 3 |
196 |
|
AY007243.1 |
- |
2, 3 |
184 |
|
AY126670.1 |
- |
3, 4 |
502 |
|
AY126671.1 |
- |
2, 3 |
328 |
|
AY126672.1 |
- |
2, 3 |
328 |
|
BC017089.2 |
- |
2, 3 |
200 |
3850358 |
BF242936.1 |
- |
|
407 |
|
View Full Product Details