Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281503.1 | NP_001268432.1 | 2 | 1538 | |
NM_052910.2 | NP_443142.1 | 1 | 1981 | ||
GenBank mRNA | AB067497.1 | - | 1 | 1980 | |
AK299479.1 | - | 1 | 1122 | ||
AY358289.1 | - | 1 | 1261 | ||
BC051738.1 | - | 1 | 1882 | 4816570 |