Gene Transcripts
- Gene Symbol
- CMTM3
- Entrez Gene ID
- 123920
- Gene Name
- CKLF like MARVEL transmembrane domain containing 3
- Gene Aliases
- BNAS2, CKLFSF3
- Chromosome Location
- Chr. 16: 66604032 - 66613892 on Build GRCh38
- UniGene ID
- Hs.298198
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144601.3 |
NP_653202.1
|
5, 6 |
821 |
|
NM_181553.2 |
NP_853531.1
|
4, 5 |
657 |
|
NR_037613.1 |
|
5, 6 |
1041 |
|
NR_037619.1 |
|
4, 5 |
714 |
|
XM_006721131.1 |
XP_006721194.1
|
5, 6 |
865 |
|
XM_011522861.1 |
XP_011521163.1
|
3, 4 |
494 |
|
GenBank mRNA |
AF479813.1 |
- |
5, 6 |
795 |
|
AK056324.1 |
- |
5, 6 |
1041 |
|
AK075075.1 |
- |
3, 4 |
354 |
|
BC023509.1 |
- |
4, 5 |
563 |
3960441 |
BC023591.2 |
- |
4, 5 |
555 |
4638047 |
BC036042.1 |
- |
4, 5 |
617 |
5271808 |
BC040227.1 |
- |
4, 5 |
608 |
5187826 |
BC047701.1 |
- |
4, 5 |
657 |
5770132 |
BI457147.1 |
- |
4, 5 |
509 |
|
CT001840.1 |
- |
4, 5 |
704 |
|
View Full Product Details