Gene Transcripts

Gene Symbol
WDSUB1
Entrez Gene ID
151525
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
UBOX6, WDSAM1
Chromosome Location
Chr. 2: 159235793 - 159286799 on Build GRCh38
UniGene ID
Hs.20848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128212.2 NP_001121684.1 11 1577
NM_001128213.1 NP_001121685.1 11 1593
NM_001307994.1 NP_001294923.1 7 1301
NM_152528.2 NP_689741.2 11 1485
XM_005246355.2 XP_005246412.2 10 1430
XM_006712341.3 XP_006712404.1 10 1065
XM_011510753.2 XP_011509055.1 11 1537
XM_011510754.2 XP_011509056.1 11 1271
XM_017003482.1 XP_016858971.1 6 1202
XM_017003483.1 XP_016858972.1 10 1392
GenBank mRNA AK093494.1 - 11 1485
AK129983.1 - 11 1593
BC029520.1 - 7 1205 5301559
BG186973.1 - 1 347
CB240898.1 - 1 342

View Full Product Details