Gene Transcripts

Gene Symbol
SASS6
Entrez Gene ID
163786
Gene Name
SAS-6 centriolar assembly protein
Gene Aliases
MCPH14, SAS-6, SAS6
Chromosome Location
Chr. 1: 100082632 - 100133095 on Build GRCh38
UniGene ID
Hs.591447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304829.1 NP_001291758.1 16 2161
NM_194292.2 NP_919268.1 17 2241
XM_017000486.1 XP_016855975.1 17 2007
GenBank mRNA AK025750.1 - 5 539
AK302513.1 - 18 1991
AK307824.1 - 16 2027
AL834265.1 - 13 1504
AY359522.1 - 17 2101
BC101026.3 - 16 1954 40007432
BX641109.1 - 16 2152
BX648406.1 - 17 1998

View Full Product Details