Gene Transcripts

Gene Symbol
TAS2R46
Entrez Gene ID
259292
Gene Name
taste 2 receptor member 46
Gene Aliases
T2R46, T2R54
Chromosome Location
Chr. 12: 11061365 - 11062294 on Build GRCh38
UniGene ID
Hs.688193
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176887.2 NP_795368.2 1 549
GenBank mRNA BC115401.2 - 1 549

View Full Product Details