Gene Transcripts

Gene Symbol
PSG11
Entrez Gene ID
5680
Gene Name
pregnancy specific beta-1-glycoprotein 11
Gene Aliases
PSBG-11, PSBG-13, PSG13, PSG14
Chromosome Location
Chr. 19: 43007657 - 43026479 on Build GRCh38
UniGene ID
Hs.741150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113410.1 NP_001106881.1 3 486
NM_002785.2 NP_002776.3 4 852
NM_203287.1 NP_976032.2 3 486
GenBank mRNA BC020711.1 - 3 476
BG571293.1 - 2 195
M69245.1 - 4 814
R66665.1 - 2 200
U25988.1 - 3 482

View Full Product Details