Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031677.3 | NP_001026847.1 | 3 | 665 | |
NM_130781.3 | NP_570137.2 | 4 | 579 | ||
NR_109789.1 | 4 | 579 | |||
NR_109790.1 | 3 | 665 | |||
GenBank mRNA | AF087904.1 | - | 3 | 383 | |
AF258583.1 | - | 3 | 409 | ||
AK021761.1 | - | 2 | 696 | ||
AK308155.1 | - | 4 | 534 | ||
AL833898.1 | - | 3 | 207 | ||
BC010006.2 | - | 4 | 502 | 4300377 | |
BC015534.1 | - | 4 | 541 | 3916832 | |
BC021263.2 | - | 3 | 625 | 4329216 | |
BT007268.1 | - | 3 | 209 |