Gene Transcripts

Gene Symbol
MAPKBP1
Entrez Gene ID
23005
Gene Name
mitogen-activated protein kinase binding protein 1
Gene Aliases
JNKBP-1, JNKBP1
Chromosome Location
Chr. 15: 41774434 - 41827855 on Build GRCh38
UniGene ID
Hs.513661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128608.1 NP_001122080.1 17, 18 2203
NM_001265611.1 NP_001252540.1 16, 17 2185
NM_014994.2 NP_055809.2 16, 17 2185
NR_049761.1 15, 16 1995
NR_049762.1 14, 15 1946
XM_006720438.1 XP_006720501.1 15, 16 2002
XM_006720439.2 XP_006720502.1 3, 4 277
XM_011521383.1 XP_011519685.1 16, 17 2020
XM_011521384.2 XP_011519686.1 17, 18 2167
XM_017022017.1 XP_016877506.1 15, 16 2296
GenBank mRNA AB011168.2 - 17, 18 2113
AB177850.1 - 8, 9 1033
AK302482.1 - 14, 15 1887
AL833267.1 - 16, 17 2105
BC036660.1 - 15, 16 2000 5266613
BC113983.1 - 16, 17 2077 40083186
BC114493.1 - 17, 18 2095 40083182
BX640787.1 - 13, 14 2350
BX648786.1 - 16, 17 2136

View Full Product Details