Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193609.1 | NP_001180538.1 | 4 | 750 | |
NM_001193610.1 | NP_001180539.1 | 4 | 750 | ||
NM_001193611.1 | NP_001180540.1 | 4 | 750 | ||
NM_001193612.1 | NP_001180541.1 | 5 | 846 | ||
NM_016261.3 | NP_057345.2 | 4 | 750 | ||
XM_005257425.1 | XP_005257482.1 | 3 | 529 | ||
XM_011524871.2 | XP_011523173.1 | 4 | 748 | ||
XM_017024716.1 | XP_016880205.1 | 4 | 748 | ||
GenBank mRNA | AB030837.1 | - | 3 | 463 | |
AB031044.1 | - | 3 | 463 | ||
AB031045.1 | - | 3 | 463 | ||
AF201333.1 | - | 3 | 462 | ||
AK022771.1 | - | 5 | 640 | ||
AK298492.1 | - | 4 | 663 | ||
AK315172.1 | - | 5 | 842 | ||
BC000258.1 | - | 4 | 725 | 3358263 |