Gene Transcripts
- Gene Symbol
- TNIP3
- Entrez Gene ID
- 79931
- Gene Name
- TNFAIP3 interacting protein 3
- Gene Aliases
- ABIN-3, LIND
- Chromosome Location
- Chr. 4: 121127161 - 121227609 on Build GRCh38
- UniGene ID
- Hs.208206
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128843.2 |
NP_001122315.2
|
5, 6 |
425 |
|
NM_001244764.1 |
NP_001231693.1
|
5, 6 |
465 |
|
NM_024873.5 |
NP_079149.3
|
2, 3 |
370 |
|
XM_011532256.2 |
XP_011530558.1
|
6, 7 |
698 |
|
XM_011532257.2 |
XP_011530559.1
|
6, 7 |
698 |
|
XM_017008619.1 |
XP_016864108.1
|
6, 7 |
698 |
|
XM_017008620.1 |
XP_016864109.1
|
6, 7 |
561 |
|
XM_017008621.1 |
XP_016864110.1
|
5, 6 |
608 |
|
XM_017008622.1 |
XP_016864111.1
|
5, 6 |
861 |
|
XM_017008623.1 |
XP_016864112.1
|
5, 6 |
471 |
|
XM_017008624.1 |
XP_016864113.1
|
4, 5 |
416 |
|
XM_017008625.1 |
XP_016864114.1
|
3, 4 |
949 |
|
GenBank mRNA |
AF277289.1 |
- |
2, 3 |
359 |
|
AJ320534.1 |
- |
2, 3 |
359 |
|
AK290409.1 |
- |
2, 3 |
351 |
|
AK301058.1 |
- |
5, 6 |
425 |
|
AK303192.1 |
- |
5, 6 |
467 |
|
BC128408.1 |
- |
2, 3 |
358 |
40115867 |
BP335455.1 |
- |
2, 3 |
370 |
|
DC389224.1 |
- |
5, 6 |
425 |
|
EF585496.1 |
- |
2, 3 |
359 |
|
View Full Product Details