Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123061038-123061539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
541332 c.71G>A p.G24E Chr.11: 123061254-123061254 on Build GRCh38
1297759 c.150G>A p.L50L Chr.11: 123061175-123061175 on Build GRCh38
1704728 c.183G>A p.M61I Chr.11: 123061142-123061142 on Build GRCh38
6257148 c.78C>A p.V26V Chr.11: 123061247-123061247 on Build GRCh38
6232515 c.189C>T p.P63P Chr.11: 123061136-123061136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904183211 Chr.11: 123061495-123061495 on Build GRCh38
rs10892958 Chr.11: 123061415-123061415 on Build GRCh38
rs923013571 Chr.11: 123061332-123061332 on Build GRCh38
rs41302367 Chr.11: 123061474-123061474 on Build GRCh38
rs201477863 Chr.11: 123061191-123061191 on Build GRCh38
rs550092655 Chr.11: 123061078-123061078 on Build GRCh38
rs117542716 Chr.11: 123061363-123061363 on Build GRCh38
rs567796127 Chr.11: 123061455-123061458 on Build GRCh38
rs767934164 Chr.11: 123061355-123061355 on Build GRCh38
rs750047710 Chr.11: 123061205-123061205 on Build GRCh38
rs144123192 Chr.11: 123061283-123061283 on Build GRCh38
rs979982971 Chr.11: 123061492-123061492 on Build GRCh38
rs770830844 Chr.11: 123061079-123061079 on Build GRCh38
rs539199549 Chr.11: 123061351-123061351 on Build GRCh38
rs959527664 Chr.11: 123061385-123061385 on Build GRCh38
rs975714996 Chr.11: 123061440-123061441 on Build GRCh38
rs766071473 Chr.11: 123061350-123061350 on Build GRCh38
rs747449685 Chr.11: 123061115-123061115 on Build GRCh38
rs770097995 Chr.11: 123061301-123061301 on Build GRCh38
rs369851462 Chr.11: 123061199-123061199 on Build GRCh38
rs753289525 Chr.11: 123061337-123061337 on Build GRCh38
rs768479550 Chr.11: 123061333-123061333 on Build GRCh38
rs895365382 Chr.11: 123061516-123061516 on Build GRCh38
rs376044760 Chr.11: 123061083-123061083 on Build GRCh38
rs758030256 Chr.11: 123061214-123061214 on Build GRCh38
rs1012327862 Chr.11: 123061395-123061395 on Build GRCh38
rs769131425 Chr.11: 123061130-123061130 on Build GRCh38
rs767193605 Chr.11: 123061163-123061163 on Build GRCh38
rs976553501 Chr.11: 123061476-123061476 on Build GRCh38
rs984037097 Chr.11: 123061455-123061455 on Build GRCh38
rs778022948 Chr.11: 123061309-123061309 on Build GRCh38
rs541230593 Chr.11: 123061439-123061439 on Build GRCh38
rs556438149 Chr.11: 123061366-123061369 on Build GRCh38
rs776953199 Chr.11: 123061075-123061075 on Build GRCh38
rs555720196 Chr.11: 123061419-123061419 on Build GRCh38
rs1027078614 Chr.11: 123061409-123061409 on Build GRCh38
rs757709713 Chr.11: 123061398-123061398 on Build GRCh38
rs201378086 Chr.11: 123061369-123061369 on Build GRCh38
rs749485084 Chr.11: 123061311-123061311 on Build GRCh38
rs760522151 Chr.11: 123061196-123061196 on Build GRCh38
rs573805467 Chr.11: 123061432-123061432 on Build GRCh38
rs750160745 Chr.11: 123061479-123061479 on Build GRCh38
rs888581357 Chr.11: 123061414-123061414 on Build GRCh38
rs749566794 Chr.11: 123061366-123061366 on Build GRCh38
rs199749364 Chr.11: 123061082-123061082 on Build GRCh38
rs747430342 Chr.11: 123061069-123061069 on Build GRCh38
rs369634495 Chr.11: 123061096-123061096 on Build GRCh38
rs753227933 Chr.11: 123061357-123061357 on Build GRCh38
rs757940184 Chr.11: 123061112-123061112 on Build GRCh38
rs771060966 Chr.11: 123061316-123061316 on Build GRCh38
rs772139172 Chr.11: 123061376-123061376 on Build GRCh38
rs760570450 Chr.11: 123061086-123061086 on Build GRCh38
rs745861967 Chr.11: 123061340-123061340 on Build GRCh38
rs773294706 Chr.11: 123061080-123061080 on Build GRCh38
rs765012279 Chr.11: 123061203-123061203 on Build GRCh38
rs113777159 Chr.11: 123061401-123061401 on Build GRCh38
rs752166960 Chr.11: 123061253-123061253 on Build GRCh38
rs756985213 Chr.11: 123061333-123061334 on Build GRCh38
rs755190153 Chr.11: 123061496-123061496 on Build GRCh38
rs75739900 Chr.11: 123061204-123061204 on Build GRCh38
rs938645828 Chr.11: 123061389-123061389 on Build GRCh38
rs778917416 Chr.11: 123061338-123061340 on Build GRCh38
rs563384300 Chr.11: 123061344-123061344 on Build GRCh38
rs745964830 Chr.11: 123061374-123061374 on Build GRCh38
rs775485056 Chr.11: 123061373-123061374 on Build GRCh38
rs373173372 Chr.11: 123061100-123061100 on Build GRCh38
rs745900938 Chr.11: 123061328-123061328 on Build GRCh38
rs755455419 Chr.11: 123061256-123061256 on Build GRCh38
rs749956024 Chr.11: 123061111-123061111 on Build GRCh38
rs549532117 Chr.11: 123061435-123061435 on Build GRCh38
rs756843449 Chr.11: 123061097-123061097 on Build GRCh38
rs1001113413 Chr.11: 123061368-123061368 on Build GRCh38
rs11551602 Chr.11: 123061231-123061231 on Build GRCh38
rs950960741 Chr.11: 123061202-123061202 on Build GRCh38
rs144664941 Chr.11: 123061332-123061333 on Build GRCh38
rs777360376 Chr.11: 123061332-123061339 on Build GRCh38
rs955710406 Chr.11: 123061456-123061456 on Build GRCh38
rs773037971 Chr.11: 123061349-123061349 on Build GRCh38
rs768851917 Chr.11: 123061071-123061071 on Build GRCh38
rs763763404 Chr.11: 123061095-123061095 on Build GRCh38
rs376156706 Chr.11: 123061172-123061172 on Build GRCh38
rs372618816 Chr.11: 123061336-123061336 on Build GRCh38
rs965626219 Chr.11: 123061420-123061420 on Build GRCh38
rs761720312 Chr.11: 123061340-123061340 on Build GRCh38
rs781396927 Chr.11: 123061160-123061160 on Build GRCh38
rs545918223 Chr.11: 123061493-123061493 on Build GRCh38
rs763047269 Chr.11: 123061180-123061180 on Build GRCh38
rs936956670 Chr.11: 123061498-123061498 on Build GRCh38
rs994330113 Chr.11: 123061073-123061073 on Build GRCh38
rs775162274 Chr.11: 123061190-123061190 on Build GRCh38
rs565779957 Chr.11: 123061347-123061347 on Build GRCh38
rs994619252 Chr.11: 123061408-123061408 on Build GRCh38
rs750885285 Chr.11: 123061421-123061421 on Build GRCh38
rs1025624159 Chr.11: 123061434-123061434 on Build GRCh38
rs758850049 Chr.11: 123061426-123061426 on Build GRCh38
rs774277451 Chr.11: 123061325-123061325 on Build GRCh38
rs754472638 Chr.11: 123061354-123061354 on Build GRCh38
rs770980780 Chr.11: 123061186-123061186 on Build GRCh38
rs927182836 Chr.11: 123061500-123061500 on Build GRCh38
rs1053965588 Chr.11: 123061504-123061504 on Build GRCh38
rs779121779 Chr.11: 123061373-123061373 on Build GRCh38
rs947249781 Chr.11: 123061360-123061360 on Build GRCh38
rs779708153 Chr.11: 123061247-123061247 on Build GRCh38
rs749193179 Chr.11: 123061064-123061064 on Build GRCh38
rs536070549 Chr.11: 123061289-123061289 on Build GRCh38
rs11823704 Chr.11: 123061470-123061470 on Build GRCh38
rs762957333 Chr.11: 123061081-123061081 on Build GRCh38
rs776774029 Chr.11: 123061485-123061485 on Build GRCh38
rs918556124 Chr.11: 123061517-123061517 on Build GRCh38
rs756515385 Chr.11: 123061361-123061361 on Build GRCh38
rs1018540488 Chr.11: 123061089-123061089 on Build GRCh38

View Full Product Details