Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31199671-31200173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_011534936.1 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5483019 c.42G>C p.A14A Chr.13: 31200126-31200126 on Build GRCh38
4965296 c.41C>A p.A14E Chr.13: 31200125-31200125 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048087156 Chr.13: 31199881-31199881 on Build GRCh38
rs898958878 Chr.13: 31200082-31200082 on Build GRCh38
rs778236602 Chr.13: 31200138-31200138 on Build GRCh38
rs987662500 Chr.13: 31200016-31200016 on Build GRCh38
rs924896071 Chr.13: 31200006-31200006 on Build GRCh38
rs912079297 Chr.13: 31200025-31200025 on Build GRCh38
rs144443154 Chr.13: 31199923-31199923 on Build GRCh38
rs940802639 Chr.13: 31199857-31199857 on Build GRCh38
rs1009988207 Chr.13: 31199950-31199950 on Build GRCh38
rs969128322 Chr.13: 31199789-31199789 on Build GRCh38
rs977720715 Chr.13: 31199999-31199999 on Build GRCh38
rs760425381 Chr.13: 31200049-31200049 on Build GRCh38
rs956274313 Chr.13: 31200009-31200009 on Build GRCh38
rs967780250 Chr.13: 31199962-31199962 on Build GRCh38
rs777497613 Chr.13: 31200108-31200125 on Build GRCh38
rs543645511 Chr.13: 31199768-31199768 on Build GRCh38
rs901022038 Chr.13: 31199945-31199945 on Build GRCh38
rs1012007377 Chr.13: 31199941-31199941 on Build GRCh38
rs1056756490 Chr.13: 31200100-31200100 on Build GRCh38
rs1034801400 Chr.13: 31199720-31199720 on Build GRCh38
rs749602953 Chr.13: 31200120-31200137 on Build GRCh38
rs946342134 Chr.13: 31199725-31199725 on Build GRCh38
rs1006748591 Chr.13: 31199730-31199730 on Build GRCh38
rs1039118339 Chr.13: 31200037-31200037 on Build GRCh38
rs764781465 Chr.13: 31200120-31200120 on Build GRCh38
rs369342759 Chr.13: 31199802-31199802 on Build GRCh38
rs1019430177 Chr.13: 31199747-31199747 on Build GRCh38
rs1047746807 Chr.13: 31200062-31200062 on Build GRCh38
rs533347362 Chr.13: 31199885-31199885 on Build GRCh38
rs1040911627 Chr.13: 31199927-31199927 on Build GRCh38
rs924864828 Chr.13: 31199746-31199746 on Build GRCh38
rs767495777 Chr.13: 31199771-31199771 on Build GRCh38
rs999173078 Chr.13: 31200140-31200140 on Build GRCh38
rs547641788 Chr.13: 31199847-31199847 on Build GRCh38
rs1030644796 Chr.13: 31200144-31200146 on Build GRCh38
rs752817236 Chr.13: 31199899-31199899 on Build GRCh38
rs900991084 Chr.13: 31199754-31199754 on Build GRCh38
rs886346534 Chr.13: 31199876-31199876 on Build GRCh38
rs759120105 Chr.13: 31200101-31200101 on Build GRCh38
rs959442980 Chr.13: 31199940-31199940 on Build GRCh38
rs943496135 Chr.13: 31200031-31200031 on Build GRCh38
rs771353044 Chr.13: 31200136-31200138 on Build GRCh38
rs753313963 Chr.13: 31200123-31200123 on Build GRCh38
rs576350779 Chr.13: 31199723-31199723 on Build GRCh38
rs890561859 Chr.13: 31199777-31199777 on Build GRCh38
rs931048411 Chr.13: 31199878-31199878 on Build GRCh38
rs1013029167 Chr.13: 31199853-31199853 on Build GRCh38
rs867643196 Chr.13: 31199874-31199874 on Build GRCh38
rs113017441 Chr.13: 31199995-31199995 on Build GRCh38
rs758969036 Chr.13: 31200132-31200132 on Build GRCh38
rs950531688 Chr.13: 31199694-31199694 on Build GRCh38
rs981937453 Chr.13: 31199711-31199711 on Build GRCh38
rs1048533225 Chr.13: 31199721-31199721 on Build GRCh38
rs576189066 Chr.13: 31199873-31199873 on Build GRCh38
rs930954435 Chr.13: 31199698-31199698 on Build GRCh38
rs1020536198 Chr.13: 31199964-31199964 on Build GRCh38
rs978985819 Chr.13: 31199744-31199744 on Build GRCh38
rs561824911 Chr.13: 31199815-31199815 on Build GRCh38
rs529218873 Chr.13: 31199819-31199819 on Build GRCh38
rs569813958 Chr.13: 31199969-31199969 on Build GRCh38
rs993054795 Chr.13: 31199846-31199846 on Build GRCh38
rs886379123 Chr.13: 31200092-31200092 on Build GRCh38
rs889432604 Chr.13: 31199890-31199890 on Build GRCh38
rs753491546 Chr.13: 31200094-31200094 on Build GRCh38
rs959017149 Chr.13: 31199843-31199843 on Build GRCh38
rs765971200 Chr.13: 31200071-31200071 on Build GRCh38

View Full Product Details